Merck
  • Simple method for the simultaneous quantification of medium-chain fatty acids and ethyl hexanoate in alcoholic beverages by gas chromatography-flame ionization detector: development of a direct injection method.
Simple method for the simultaneous quantification of medium-chain fatty acids and ethyl hexanoate in alcoholic beverages by gas chromatography-flame ionization detector: development of a direct injection method.
Journal of chromatography. A (2011-09-20)
Kei Takahashi, Nami Goto-Yamamoto
ABSTRACT
Free medium-chain fatty acids (MCFAs) can negatively influence the fermentation process and taste quality in alcoholic beverages. Ethyl hexanoate is important in providing a fruit-like flavour to drinks, particularly in Japanese sake. In this study, we developed a direct injection method for a gas chromatography-flame ionization detector following the semi-purification of chemical components, such as esters, alcohols and MCFAs in alcoholic beverages. Evaluation of MCFAs by this method gave a limit of detection on the order of sub-ppm and relative standard deviations less than 10% in standard solution. Good repeatability and recovery rates against MCFAs and ethyl hexanoate were also obtained in non-distilled real alcoholic beverages. Because this method enabled us to simultaneously quantify the concentrations of MCFAs and ethyl hexanoate, the proportion of ester against MCFAs was proposed as a quality control index. This method could be suitable for routine analysis in the alcohol beverage industry.
Sigma-Aldrich
Ethyl hexanoate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl hexanoate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl hexanoate, ≥99%