Merck
[GC-MS analysis of volatile oil of olibanum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2011-08-04)
Jinfeng Zhao, Chunlan Zhou, Fengqin Zhou, Taiming Wei
ABSTRACT
To analyze the volatile oil of olibanum and apply scientific evidences for its applications. The volatile oil was analyzed by GC-MS. One hundred components were identified,accounting for 91.26% of the total volatile oil, and the main components were octyl acetate, beta-elemene. It contains some transdermal absorption enhancers. The components of olibanum volatile oil were complicated; the connatural transdermal absorption enhancers make it possible to use in external preparation.
Sigma-Aldrich
Octyl acetate, ≥99%
Sigma-Aldrich
Octyl acetate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Octyl acetate, ≥98%, FCC, FG