Merck
  • Direction-selective circuitry in rat retina develops independently of GABAergic, cholinergic and action potential activity.
Direction-selective circuitry in rat retina develops independently of GABAergic, cholinergic and action potential activity.
PloS one (2011-05-17)
Le Sun, Xu Han, Shigang He
ABSTRACT
The ON-OFF direction selective ganglion cells (DSGCs) in the mammalian retina code image motion by responding much more strongly to movement in one direction. They do so by receiving inhibitory inputs selectively from a particular sector of processes of the overlapping starburst amacrine cells, a type of retinal interneuron. The mechanisms of establishment and regulation of this selective connection are unknown. Here, we report that in the rat retina, the morphology, physiology of the ON-OFF DSGCs and the circuitry for coding motion directions develop normally with pharmacological blockade of GABAergic, cholinergic activity and/or action potentials for over two weeks from birth. With recent results demonstrating light independent formation of the retinal DS circuitry, our results strongly suggest the formation of the circuitry, i.e., the connections between the second and third order neurons in the visual system, can be genetically programmed, although emergence of direction selectivity in the visual cortex appears to require visual experience.
Sigma-Aldrich
Cesium methanesulfonate, used in patch clamp techniques
Millipore
MCE Membrane Filter, 0.8 μm Pore Size, MF-Millipore, filter diam. 25 mm, hydrophilic, black
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)