Merck
  • Diaxial diureido decalins as compact, efficient, and tunable anion transporters.
Diaxial diureido decalins as compact, efficient, and tunable anion transporters.
Journal of the American Chemical Society (2011-01-25)
Sabir Hussain, Peter R Brotherhood, Luke W Judd, Anthony P Davis
ABSTRACT
Decalins bearing two axial -NHCONHAr substituents and an ester-linked alkyl side chain have been synthesized and studied as anion receptors and transporters. The design relates to steroid-based "cholapods" but is more compact and less intrinsically lipophilic. Transport rates depend on both NHAr and the alkyl side chain. High activities can be achieved; with optimal substitution, chloride-nitrate exchange across vesicle membranes is measurable at transporter/lipid ratios as low as 1:250,000.
Sigma-Aldrich
cis-Decahydronaphthalene, 99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%
Sigma-Aldrich
trans-Decahydronaphthalene, 99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%