Merck
  • A tetrahydrofuranol from the leaves of Michelia compressa var. lanyuensis (Magnoliaceae).
A tetrahydrofuranol from the leaves of Michelia compressa var. lanyuensis (Magnoliaceae).
Natural product research (2010-11-26)
Chung-Yi Chen
ABSTRACT
Pressalanin A, a 2-functionalized tetrahydrofuranol, has been isolated from the leaves of Michelia compressa var. lanyuensis (Magnoliaceae), and the structure was determined on the basis of a spectroscopic analysis.
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 98%
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, ≥98%
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 99%