Merck
  • CuI-catalyzed amination of arylhalides with guanidines or amidines: a facile synthesis of 1-H-2-substituted benzimidazoles.
CuI-catalyzed amination of arylhalides with guanidines or amidines: a facile synthesis of 1-H-2-substituted benzimidazoles.
The Journal of organic chemistry (2009-06-17)
Xiaohu Deng, Heather McAllister, Neelakandha S Mani
ABSTRACT
CuI/L5 (N,N'-dimethylethylenediamine) proves to be an efficient catalyst system for the amination of arylhalides with guanidines. The same catalyst system is then successfully applied to the one-step synthesis of 1-H-2-amino-benzimidazoles through tandem aminations of 1,2-dihaloarenes in modest yields. This methodology is also applicable for the preparation of 1-H or 1-substutituted 2-aryl- or 2-alkyl-benzimidazoles.
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 85%
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 98%