Merck
High volume image guided injections in chronic Achilles tendinopathy.
Disability and rehabilitation (2008-07-09)
Otto Chan, Dominic O'Dowd, Nat Padhiar, Dylan Morrissey, John King, Rosy Jalan, Nicola Maffulli, Tom Crisp
ABSTRACT
To determine the effectiveness of high volume image guided injections (HVIGI) for chronic Achilles tendinopathy. We included in the study 30 consecutive patients (mean age 37.2 years, range 24 - 58 years) with Achilles tendinopathy for a mean of 35.8 months (range 2 - 276 months) who had failed to improve after a three-month programme of eccentric loading of the gastro-soleus complex. Patients were injected with 10 ml of 0.5% Bupivacaine Hydrochloride, 25 mg Hydrocortisone acetate, and up to 40 ml of injectable normal saline. A study-specific questionnaire and the Victorian Institute of Sport Assessment - Achilles tendon (VISA-A) were retrospectively administered to assess short- and long-term pain and functional improvement. Some 21 patients (70%) responded. Patients reported significant short-term improvement at 4 weeks of both pain (mean change 50 mm, [SD 28, p < 0.0001], from a mean of 76 mm [SD 18.2], to a mean of 25 mm [SD 23.3]), and function scores (mean change 51 mm, [SD 31.2, p < 0.0001], from a mean of 78 mm [SD 20.8], to a mean of 27 mm [SD 28.4]). Patients also reported significant long-term improvement in symptoms using the VISA-A questionnaire (mean change 31.2 points, [SD = 28, p < 0.0001], from a mean of 44.8 points [SD 17.7], to a mean of 76.2 points [SD 24.6]) at a mean of 30.3 weeks from the injection. HVIGI significantly reduces pain and improves function in patients with resistant Achilles tendinopathy in the short- and long-term.
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Hydrocortisone acetate, European Pharmacopoeia (EP) Reference Standard