Merck
  • Domino reaction of 3-(2-formylphenoxy)propenoates and amines: a novel synthesis of 1,4-dihydropyridines from salicaldehydes, ethyl propiolate, and amines.
Domino reaction of 3-(2-formylphenoxy)propenoates and amines: a novel synthesis of 1,4-dihydropyridines from salicaldehydes, ethyl propiolate, and amines.
The Journal of organic chemistry (2007-09-11)
Sun-Liang Cui, Jun Wang, Xu-Feng Lin, Yan-Guang Wang
ABSTRACT
A novel synthesis of Hantzsch-type N-substituted 1,4-dihydropyridines from salicaldehydes, ethyl propiolate, and amines has been developed. Salicaldehydes were treated with ethyl propiolate in the presence of N-methylmorpholine to give ethyl 3-(2-formylphenoxy)propenoates. Three equivalents of ethyl 3-(2-formylphenoxy)propenoates reacted with 1 equiv of amines under trifluoroacetic acid (TFA) catalyst to furnish the corresponding N-substituted 1,4-dihydropyridines in good to excellent yields, recovering the starting material salicaldehydes. A possible mechanism for the domino process was proposed. Furthermore, the products can be easily derived via further transformations and three of them exhibited strong fluorescence (Phif = 0.36-0.63).
Sigma-Aldrich
Ethyl propionate, 99%
Sigma-Aldrich
Ethyl propionate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Ethyl propionate, ≥97%, FCC, FG