Merck
  • Analysis of dencichine in Panax notoginseng by gas chromatography-mass spectrometry with ethyl chloroformate derivatization.
Analysis of dencichine in Panax notoginseng by gas chromatography-mass spectrometry with ethyl chloroformate derivatization.
Journal of pharmaceutical and biomedical analysis (2006-10-13)
Guo-Xiang Xie, Yun-Ping Qiu, Ming-Feng Qiu, Xian-Fu Gao, Yu-Min Liu, Wei Jia
ABSTRACT
Dencichine (beta-N-oxalyl-l-alpha,beta-diaminopropionic acid) is a haemostatic agent present in well-known traditional Chinese medicinal herbs such as Panax notoginseng, as well as other Panax species. It is also a reported neurotoxic agent found in Lathyrus sativus (grass pea seed) and cycad seeds. A method was developed for quantitative determination of the non-protein amino acid, dencichine, in plant samples of P. notoginseng and the adventitious roots directly from the explants of P. notoginseng after derivatization with ethyl chloroformate (ECF) by gas chromatography-mass spectrometry (GC-MS). l-2-chlorophenylalanine was used as an internal standard. Calibration curves were linear (r(2)=0.9988, n=6) in the range of 10-800 microg/ml for dencichine. Limit of detection and quantification for dencichine were 0.5 microg/ml and 2 microg/ml, respectively. This rapid and specific method may be applied to the quantification of dencichine in complex medicinal plants and their products.
Sigma-Aldrich
Ethyl chloroformate, 97%
Sigma-Aldrich
Ethyl chloroformate, purum, ≥98.0% (GC)