Merck
  • Changes of the volatile profile and artifact formation in Daidai (Citrus aurantium) cold-pressed peel oil on storage.
Changes of the volatile profile and artifact formation in Daidai (Citrus aurantium) cold-pressed peel oil on storage.
Journal of agricultural and food chemistry (2003-06-26)
Simon Muhoho Njoroge, Hiroyuki Ukeda, Masayoshi Sawamura
ABSTRACT
The chemical changes and artifact formation in daidai (Citrus aurantium L. var. Cyathifera Y. Tanaka) cold-pressed peel oil upon storage at 20, 5, and -21 degrees C for 3, 6, and 12 months were investigated using capillary gas chromatography (GC) and GC-MS. Major changes occurred in the oil stored at 20 and 5 degrees C. No changes were found at -21 degrees C. Monoterpene hydrocarbons decreased from 98.0 to 66.4% upon 12 months at 20 degrees C, while sesquiterpene hydrocarbons and alcohols increased from 0.1 to 2.4% and from 0.3 to 7.9%, respectively. Notable decreases of germacrene D, myrcene, linalyl acetate, and limonene occurred. Prominent increases of cis-carveol, trans-beta-farnesene, trans-p-2,8-menthadien-1-ol, linalool, and beta-caryophyllene were found. Thirty-four artifact compounds constituting 17.0% of the total volatile compounds were formed upon 12 months at 20 degrees C. The artifacts consisted of 13 alcohols (6.0%), five carbonyl compounds (5.3%), seven esters (4.9%), three epoxides (0.4%), four hydrocarbons (0.3%), and two unidentified. The prominent artifact compounds were (+)-carvone, trans,trans-farnesyl acetate, sabinene hydrate, 1-octen-3-ol, cis,cis-farnesyl acetate, and dihydrocarveol acetate. The results could be applied for monitoring and control of the flavor quality of daidai essential oil and related products.
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG