Merck
  • Macrophage are the principal reservoir and sustain high virus loads in rhesus macaques after the depletion of CD4+ T cells by a highly pathogenic simian immunodeficiency virus/HIV type 1 chimera (SHIV): Implications for HIV-1 infections of humans.
Macrophage are the principal reservoir and sustain high virus loads in rhesus macaques after the depletion of CD4+ T cells by a highly pathogenic simian immunodeficiency virus/HIV type 1 chimera (SHIV): Implications for HIV-1 infections of humans.
Proceedings of the National Academy of Sciences of the United States of America (2001-01-03)
T Igarashi, C R Brown, Y Endo, A Buckler-White, R Plishka, N Bischofberger, V Hirsch, M A Martin
ABSTRACT
The highly pathogenic simian immunodeficiency virus/HIV type 1 (SHIV) chimeric virus SHIV(DH12R) induces a systemic depletion of CD4(+) T lymphocytes in rhesus monkeys during the initial 3-4 weeks of infection. Nonetheless, high levels of viral RNA production continue unabated for an additional 2-5 months. In situ hybridization and immunohistochemical analyses revealed that tissue macrophage in the lymph nodes, spleen, gastrointestinal tract, liver, and kidney sustain high plasma virus loads in the absence of CD4(+) T cells. Quantitative confocal immunofluorescence analysis indicated that greater than 95% of the virus-producing cells in these tissues are macrophage and less than 2% are T lymphocytes. Interestingly, the administration of a potent reverse transcriptase inhibitor blocked virus production during the early T cell phase but not during the later macrophage phase of the SHIV(DH12R) infection. When interpreted in the context of HIV-1 infections, these results implicate tissue macrophage as an important reservoir of virus in vivo. They become infected during the acute infection, gradually increase in number over time, and can be a major contributor to total body virus burden during the symptomatic phase of the human infection.
Sigma-Aldrich
Pinacolyl methylphosphonate, 97%