Merck
  • Syntheses of stereochemically restricted lactone-type analogues of jasmonic acids.
Syntheses of stereochemically restricted lactone-type analogues of jasmonic acids.
Bioscience, biotechnology, and biochemistry (2000-10-31)
H Toshima, H Aramaki, A Ichihara
ABSTRACT
5-Oxa-7-epi-jasmonic acid and 5-oxa-jasmonic acid, which are stereochemically restricted lactone-type analogues of jasmonic acids, were synthesized via three-component coupling of 2(5H)-furanone, tert-butyl acetate and 1-bromo-2-pentyne. After acidic deprotection of the tert-butyl esters, the (Z)-olefin was introduced by catalytic partial reduction with the Lindlar catalyst to give the desired analogues.
Sigma-Aldrich
tert-Butyl acetate, ≥99%
Sigma-Aldrich
1-Bromo-2-pentyne, 97%