Merck
  • Salt-bridge dynamics control substrate-induced conformational change in the membrane transporter GlpT.
Salt-bridge dynamics control substrate-induced conformational change in the membrane transporter GlpT.
Journal of molecular biology (2008-04-09)
Christopher J Law, Jonas Almqvist, Adam Bernstein, Regina M Goetz, Yafei Huang, Celine Soudant, Aatto Laaksonen, Sven Hovmöller, Da-Neng Wang
ABSTRACT
Active transport of substrates across cytoplasmic membranes is of great physiological, medical and pharmaceutical importance. The glycerol-3-phosphate (G3P) transporter (GlpT) of the E. coli inner membrane is a secondary active antiporter from the ubiquitous major facilitator superfamily that couples the import of G3P to the efflux of inorganic phosphate (P(i)) down its concentration gradient. Integrating information from a novel combination of structural, molecular dynamics simulations and biochemical studies, we identify the residues involved directly in binding of substrate to the inward-facing conformation of GlpT, thus defining the structural basis for the substrate-specificity of this transporter. The substrate binding mechanism involves protonation of a histidine residue at the binding site. Furthermore, our data suggest that the formation and breaking of inter- and intradomain salt bridges control the conformational change of the transporter that accompanies substrate translocation across the membrane. The mechanism we propose may be a paradigm for organophosphate:phosphate antiporters.
Sigma-Aldrich
sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt, ≥93% (GC)
Pricing and availability is not currently available.