Merck
  • Glucose oxidation-dependent survival of activated B cells provides a putative novel therapeutic target for lupus treatment.
Glucose oxidation-dependent survival of activated B cells provides a putative novel therapeutic target for lupus treatment.
iScience (2023-08-28)
John J Wilson, Jian Wei, Andrea R Daamen, John D Sears, Elaine Bechtel, Colleen L Mayberry, Grace A Stafford, Lesley Bechtold, Amrie C Grammer, Peter E Lipsky, Derry C Roopenian, Chih-Hao Chang
ABSTRACT
Aberrant metabolic demand is observed in immune/inflammatory disorders, yet the role in pathogenesis remains unclear. Here, we discover that in lupus, activated B cells, including germinal center B (GCB) cells, have remarkably high glycolytic requirement for survival over T cell populations, as demonstrated by increased metabolic activity in lupus-activated B cells compared to immunization-induced cells. The augmented reliance on glucose oxidation makes GCB cells vulnerable to mitochondrial ROS-induced oxidative stress and apoptosis. Short-term glycolysis inhibition selectively reduces pathogenic activated B in lupus-prone mice, extending their lifespan, without affecting T follicular helper cells. Particularly, BCMA-expressing GCB cells rely heavily on glucose oxidation. Depleting BCMA-expressing activated B cells with APRIL-based CAR-T cells significantly prolongs the lifespan of mice with severe autoimmune disease. These results reveal that glycolysis-dependent activated B and GCB cells, especially those expressing BCMA, are potentially key lupus mediators, and could be targeted to improve disease outcomes.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Rotenone, A mitochondrial toxin and a potent, reversible, and competitive inhibitor of complex I (NADH-CoQ reductase) of the respiratory chain.
Sigma-Aldrich
Antimycin A from Streptomyces sp.