Merck
  • Two-dimensional liquid chromatography coupled to tandem mass spectrometry for vitamin D metabolite profiling including the C3-epimer-25-monohydroxyvitamin D3.
Two-dimensional liquid chromatography coupled to tandem mass spectrometry for vitamin D metabolite profiling including the C3-epimer-25-monohydroxyvitamin D3.
Journal of chromatography. A (2016-05-18)
A Mena-Bravo, F Priego-Capote, M D Luque de Castro
ABSTRACT
A method based on automated on-line solid phase extraction coupled to two-dimensional liquid chromatography with tandem mass spectrometry detection (SPE-2DLC-MS/MS) is here reported for vitamin D metabolite profiling in human serum with absolute quantitation. Two-dimensional LC was configured with two complementary analytical columns, pentafluorophenyl (PFP) and C18 phases, for determination of 25 hydroxyvitamin D3 epimers and the resting bioactive metabolites of vitamin D (D3 and D2)-25-hydroxyvitamin D2, 1,25-dihydroxyvitamin D3, 1,25-dihydroxyvitamin D2 and 24,25-dihydroxyvitamin D3. Quantitative determination was supported on the use of a stable isotopic labelled internal standard for each analyte and the resulting method was validated by analysis of a standard reference material certified by the National Institute of Standards & Technology (NIST-972a) and 5 samples provided by the vitamin D External Quality Assurance Scheme (DEQAS). The limits of detection were between 9 and 90pg/mL for the eight analytes, and precision, expressed as relative standard deviation, was lower than 11.6%. Two-dimensional LC has shown to be the key to discriminate between 25 hydroxyvitamin D3 epimers in a quantitative analysis also involving dihydroxyvitamin D metabolites.
Product Description
Vitamin D metabolites in frozen human serum, NIST® SRM® 972a