Merck
  • Simultaneous measurement of 13 circulating vitamin D3 and D2 mono and dihydroxy metabolites using liquid chromatography mass spectrometry.
Simultaneous measurement of 13 circulating vitamin D3 and D2 mono and dihydroxy metabolites using liquid chromatography mass spectrometry.
Clinical chemistry and laboratory medicine (2021-05-21)
Carl Jenkinson, Reena Desai, Andrzej T Slominski, Robert C Tuckey, Martin Hewison, David J Handelsman
ABSTRACT
Clinical evaluation of vitamin D status is conventionally performed by measuring serum levels of a single vitamin D metabolite, 25-hydroxyvitamin D predominantly by immunoassay methodology. However, this neglects the complex metabolic pathways involved in vitamin D bioactivity, including two canonical forms D3 and D2, bioactive 1,25-dihydroxy metabolites and inactive 24-hydroxy and other metabolites. Liquid chromatography-tandem mass spectrometry (LC-MS/MS) can measure multiple analytes in a sample during a single run with high sensitivity and reference level specificity. We therefore aimed to develop and validate a LC-MS/MS method to measure simultaneously 13 circulating vitamin D metabolites and apply it to 103 human serum samples. The LC-MS/MS method using a Cookson-type derivatization reagent phenyl-1,2,4-triazoline-3,5-dione (PTAD) quantifies 13 vitamin D metabolites, including mono and dihydroxy-metabolites, as well as CYP11A1-derived D3 and D2 metabolites in a single run. The lower limit of quantitation was 12.5 pg/mL for 1,25(OH)2D3 with accuracy verified by analysis of National Institute of Standards and Technology (NIST) 972a standards. Quantification of seven metabolites (25(OH)D3, 25(OH)D2, 3-epi-25(OH)D3, 20(OH)D3, 24,25(OH)2D3, 1,25(OH)2D3 and 1,20S(OH)2D3) was consistently achieved in human serum samples. This profiling method can provide new insight into circulating vitamin D metabolite pathways forming the basis for improved understanding of the role of vitamin D in health and disease.
Product Description
Vitamin D metabolites in frozen human serum, NIST® SRM® 972a