Merck
Ultrasensitive negative capacitance phototransistors.
Nature communications (2020-01-05)
Luqi Tu, Rongrong Cao, Xudong Wang, Yan Chen, Shuaiqin Wu, Fang Wang, Zhen Wang, Hong Shen, Tie Lin, Peng Zhou, Xiangjian Meng, Weida Hu, Qi Liu, Jianlu Wang, Ming Liu, Junhao Chu
ABSTRACT
Sensitive photodetection is crucial for modern optoelectronic technology. Two-dimensional molybdenum disulfide (MoS2) with unique crystal structure, and extraordinary electrical and optical properties is a promising candidate for ultrasensitive photodetection. Previously reported methods to improve the performance of MoS2 photodetectors have focused on complex hybrid systems in which leakage paths and dark currents inevitably increase, thereby reducing the photodetectivity. Here, we report an ultrasensitive negative capacitance (NC) MoS2 phototransistor with a layer of ferroelectric hafnium zirconium oxide film in the gate dielectric stack. The prototype photodetectors demonstrate a hysteresis-free ultra-steep subthreshold slope of 17.64 mV/dec and ultrahigh photodetectivity of 4.75 × 1014 cm Hz1/2 W-1 at room temperature. The enhanced performance benefits from the combined action of the strong photogating effect induced by ferroelectric local electrostatic field and the voltage amplification based on ferroelectric NC effect. These results address the key challenges for MoS2 photodetectors and offer inspiration for the development of other optoelectronic devices.
Sigma-Aldrich
Tetrakis(ethylmethylamido)hafnium(IV), packaged for use in deposition systems
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 kg
Please contact Customer Service for Availability
€80.10
To order products, please contact your local dealer.
Sigma-Aldrich
Tetrakis(ethylmethylamido)hafnium(IV), ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 kg
Please contact Customer Service for Availability
€80.10
To order products, please contact your local dealer.