Merck
  • Synthesis of Fluorescent Dansyl Derivatives of Methoxyamine and Diphenylhydrazine as Free Radical Precursors.
Synthesis of Fluorescent Dansyl Derivatives of Methoxyamine and Diphenylhydrazine as Free Radical Precursors.
International journal of molecular sciences (2020-05-24)
Bianca Patrascu, Sorin Mocanu, Anca Coman, Augustin M Madalan, Codruta Popescu, Anca Paun, Mihaela Matache, Petre Ionita
ABSTRACT
Starting from dansyl-chloride, in reaction with 1,1-diphenylhydrazine and methoxyamine, two new fluorescent derivatives 1 and 2 were obtained and characterized by NMR, IR, UV-Vis, HR-MS, and fluorescence spectroscopy. The single-crystal X-ray structure was obtained for compound 2. Both compounds generate free radicals by oxidation, as demonstrated by ESR spectroscopy. Compound 1 generates the corresponding hydrazyl-persistent free radical, evidenced directly by ESR spectroscopy, while compound 2 generates in the first instance the methoxyaminyl short-lived free radical, which decomposes rapidly with the formation of the methoxy radical, evidenced by the ESR spin-trapping technique. By oxidation of compounds 1 and 2, their fluorescence is quenched.
Sigma-Aldrich
Endothelial Cell Growth Medium MV, SupplementPack containing all media supplements as individual vials, 1 Pack for 500 ml
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)