Merck
  • Modeling alpha-synuclein pathology in a human brain-chip to assess blood-brain barrier disruption.
Modeling alpha-synuclein pathology in a human brain-chip to assess blood-brain barrier disruption.
Nature communications (2021-10-10)
Iosif Pediaditakis, Konstantia R Kodella, Dimitris V Manatakis, Christopher Y Le, Chris D Hinojosa, William Tien-Street, Elias S Manolakos, Kostas Vekrellis, Geraldine A Hamilton, Lorna Ewart, Lee L Rubin, Katia Karalis
ABSTRACT
Parkinson's disease and related synucleinopathies are characterized by the abnormal accumulation of alpha-synuclein aggregates, loss of dopaminergic neurons, and gliosis of the substantia nigra. Although clinical evidence and in vitro studies indicate disruption of the Blood-Brain Barrier in Parkinson's disease, the mechanisms mediating the endothelial dysfunction is not well understood. Here we leveraged the Organs-on-Chips technology to develop a human Brain-Chip representative of the substantia nigra area of the brain containing dopaminergic neurons, astrocytes, microglia, pericytes, and microvascular brain endothelial cells, cultured under fluid flow. Our αSyn fibril-induced model was capable of reproducing several key aspects of Parkinson's disease, including accumulation of phosphorylated αSyn (pSer129-αSyn), mitochondrial impairment, neuroinflammation, and compromised barrier function. This model may enable research into the dynamics of cell-cell interactions in human synucleinopathies and serve as a testing platform for target identification and validation of novel therapeutics.
Sigma-Aldrich
Monoclonal Anti-Tyrosine Hydroxylase antibody produced in mouse, clone TH-16, ascites fluid
Sigma-Aldrich
Anti-LMX-1 Antibody, clone 4L12, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets