Merck
  • An ethylene biosynthesis enzyme controls quantitative variation in maize ear length and kernel yield.
An ethylene biosynthesis enzyme controls quantitative variation in maize ear length and kernel yield.
Nature communications (2021-10-07)
Qiang Ning, Yinan Jian, Yanfang Du, Yunfu Li, Xiaomeng Shen, Haitao Jia, Ran Zhao, Jimin Zhan, Fang Yang, David Jackson, Lei Liu, Zuxin Zhang
ABSTRACT
Maize ear size and kernel number differ among lines, however, little is known about the molecular basis of ear length and its impact on kernel number. Here, we characterize a quantitative trait locus, qEL7, to identify a maize gene controlling ear length, flower number and fertility. qEL7 encodes 1-aminocyclopropane-1- carboxylate oxidase2 (ACO2), a gene that functions in the final step of ethylene biosynthesis and is expressed in specific domains in developing inflorescences. Confirmation of qEL7 by gene editing of ZmACO2 leads to a reduction in ethylene production in developing ears, and promotes meristem and flower development, resulting in a ~13.4% increase in grain yield per ear in hybrids lines. Our findings suggest that ethylene serves as a key signal in inflorescence development, affecting spikelet number, floral fertility, ear length and kernel number, and also provide a tool to improve grain productivity by optimizing ethylene levels in maize or in other cereals.
Product Description
Roche
Ribonucleoside Triphosphate Set, pkg of 4 × 200 μL (4 x 20 μmol; 100 mM, each), suitable for DNA sequencing, >98% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep