Merck
  • Identification of glutathionyl-3-hydroxykynurenine glucoside as a novel fluorophore associated with aging of the human lens.
Identification of glutathionyl-3-hydroxykynurenine glucoside as a novel fluorophore associated with aging of the human lens.
The Journal of biological chemistry (1999-07-20)
B Garner, S Vazquez, R Griffith, R A Lindner, J A Carver, R J Truscott
ABSTRACT
A novel fluorophore was isolated from human lenses using high performance liquid chromatography (HPLC). The new fluorophore was well separated from 3-hydroxykynurenine glucoside (3-OHKG) and its deaminated isoform, 4-(2-amino-3-hydroxyphenyl)-4-oxobutanoic acid O-glucoside, which are known UV filter compounds. The new compound exhibited UV absorbance maxima at 260 and 365 nm, was fluorescent (Ex(360 nm)/Em(500 nm)), and increased in concentration with age. Further analysis of the purified compound by microbore HPLC with in-line electrospray ionization mass spectrometry revealed a molecular mass of 676 Da. This mass corresponds to that of an adduct of GSH with a deaminated form of 3-OHKG. This adduct was synthesized using 3-OHKG and GSH as starting materials. The synthetic glutathionyl-3-hydroxykynurenine glucoside (GSH-3-OHKG) adduct had the same HPLC elution time, thin-layer chromatography R(F) value, UV absorbance maxima, fluorescence characteristics, and mass spectrum as the lens-derived fluorophore. Furthermore, the (1)H and (13)C NMR spectra of the synthetic adduct were entirely consistent with the proposed structure of GSH-3-OHKG. These data indicate that GSH-3-OHKG is present as a novel fluorophore in aged human lenses. The GSH-3-OHKG adduct was found to be less reactive with beta-glucosidase compared with 3-OHKG, and this could be due to a folded conformation of the adduct that was suggested by molecular modeling.
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, ≥2 units/mg solid