Merck
  • Quercetin Alleviates Oxidative Damage by Activating Nuclear Factor Erythroid 2-Related Factor 2 Signaling in Porcine Enterocytes.
Quercetin Alleviates Oxidative Damage by Activating Nuclear Factor Erythroid 2-Related Factor 2 Signaling in Porcine Enterocytes.
Nutrients (2021-02-04)
Hai Jia, Yunchang Zhang, Xuemeng Si, Yuhang Jin, Da Jiang, Zhaolai Dai, Zhenlong Wu
ABSTRACT
Oxidative stress has been implicated in the etiology of multiple gastrointestinal disorders, such as irritable bowel syndrome and inflammatory bowel disease. This study was conducted to evaluate effects of natural product quercetin on diquat-induced oxidative stress in porcine enterocytes and underlying mechanisms. Intestinal porcine epithelial cell line 1 (IPEC-1) cells pretreated with or without quercetin (5 μM, 24 h) were incubated with vehicle or diquat (100 μM) for 6 h. The results showed that diquat treatment induced apoptosis in a caspase-3-dependent manner, as accompanied by elevated reactive oxygen species (ROS) production, increased mitochondrial depolarization, and reduced the abundance of tight junction proteins. These adverse effects of diquat were remarkably abrogated by quercetin administration. Further study indicated that the protective effect of quercetin was associated with elevated protein abundance of nuclear factor erythroid 2-related factor 2 (Nrf2) and increased intracellular glutathione (GSH) content. Interestingly, the beneficial effects of quercetin on diquat-induced oxidative damage were abolished by all-trans-retinoic acid (Atra), a specific inhibitor of Nrf2, indicating a Nrf2-dependent regulation manner. The results show that quercetin attenuates diquat-induced cell injury by promoting protein abundance of Nrf2 and regulating GSH-related redox homeostasis in enterocytes. These findings provide new insights into a function role of quercetin in maintaining intestinal homeostasis.
Product Description
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diquat dibromide monohydrate, PESTANAL®, analytical standard
Sigma-Aldrich
JC-1, powder or solid (Crystals)