Merck
  • The Type and Source of Reactive Oxygen Species Influences the Outcome of Oxidative Stress in Cultured Cells.
The Type and Source of Reactive Oxygen Species Influences the Outcome of Oxidative Stress in Cultured Cells.
Cells (2021-05-06)
Steffi Goffart, Petra Tikkanen, Craig Michell, Trevor Wilson, Jaakko L O L O Pohjoismäki
ABSTRACT
Oxidative stress can be modeled using various different experimental approaches, such as exposing the cells or organisms to oxidative chemicals. However, the actual effects of these chemicals, outside of the immediate measured effect, have attracted relatively little attention. We show here that three commonly used oxidants, menadione, potassium bromate, and hydrogen peroxide, while known to function differently, also elicit different types of responses in HEK293T cells. Menadione and bromate exposure mainly trigger an integrated stress response, whereas hydrogen peroxide affects cellular processes more diversely. Interestingly, acute oxidative stress does not universally cause notable induction of DNA repair or antioxidant defense mechanisms. We also provide evidence that cells with previous experience of oxidative stress show adaptive changes in their responses when the stress is renewed. Our results urge caution when comparing studies where different sources of oxidative stress have been used or when generalizing the findings of these studies to other oxidant types or tissues.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Vdac1 antibody produced in mouse, clone S152B-23, purified immunoglobulin