Merck
  • Design, synthesis and cytotoxic properties of novel 1-[4-(2-alkylaminoethoxy)phenylcarbonyl]-3,5-bis(arylidene)-4-piperidones and related compounds.
Design, synthesis and cytotoxic properties of novel 1-[4-(2-alkylaminoethoxy)phenylcarbonyl]-3,5-bis(arylidene)-4-piperidones and related compounds.
European journal of medicinal chemistry (2006-09-26)
Umashankar Das, Jane Alcorn, Anuraag Shrivastav, Rajendra K Sharma, Erik De Clercq, Jan Balzarini, Jonathan R Dimmock
ABSTRACT
The 3,5-bis(arylidene)-4-piperidones 1 contain the 1,5-diaryl-3-oxo-1,4-pentadienyl pharmacophore which is considered to interact at a complementary binding site in susceptible neoplasms. The hypothesis was formulated that the presence of an acyl group attached to the piperidyl nitrogen atom in series 1 may interact with an additional binding site thereby enhancing cytotoxic potencies. This concept led to the synthesis of various N-acyl-3,5-bis(arylidene)-4-piperidones 3-7 many of which displayed significant cytotoxicity towards a variety of cancer cell lines. A comparison of the potencies between the compounds in series 1 and the related nonquaternary analogues 3-6 revealed that in approximately half of the comparisons made, the N-acyl analogues had increased potencies.
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Melphalan, powder
Pricing and availability is not currently available.