Merck
  • A sensitive liquid chromatography-tandem mass spectrometric method for determination of five β-blockers after labeling with either hydrazonoyl chloride or dansyl chloride reagent.
A sensitive liquid chromatography-tandem mass spectrometric method for determination of five β-blockers after labeling with either hydrazonoyl chloride or dansyl chloride reagent.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2020-09-20)
Alaa Khedr, Ahdab N Khayyat, Abdel-Nasser A El-Shorbagi, Ahmed K Kammoun
ABSTRACT
A sensitive liquid chromatography-tandem mass spectrometry (LC-MS) method was developed for the screening of five β-blockers (BBs), including atenolol, metoprolol, bisoprolol, propranolol, and betaxolol, in rabbit plasma. An inhouse prepared hydrazonoyl chloride compound (UOSA54) and dansyl chloride (DNS) were successfully coupled with BBs via the amino functional group and analyzed by LC-MS/MS. The excess hydrazonoyl chloride was characterized by a negligible ionization suppression at the electrospray-ionization (ESI) source, which enables the analysis of BBs at a low concentration level. The relative ESI-MS response of derivatized to underivatized BBs was enhanced 2.3 to 3.7-fold. The developed method could be applied for screening of BBs in samples by searching the most abundant MS product ions, including m/z 169 and 211, in addition to the precursor ion and the cleavage of ether moiety. The method could be applied for trace analysis and screening of BBs abuse. The use of UOSA54 was adventitious over dansylated derivatives because of minimal reaction by-products and the negligible ionization suppression effect. The extraction efficiency of BBs from rabbit plasma was improved to reach 77.5-93.9% using tert-butylamine and Chromabond® C18ec-100 mg column. The optimal reaction conditions were optimized and validated. The linear range of analyzed BBs in rabbit plasma was within the range of 0.1 to 25.0 ng/mL, while the limit of quantification (LO Q) was ranged from 0.10 to 0.25 ng/mL.
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bisoprolol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Betaxolol hydrochloride, >98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Naphthalenesulfonyl chloride, 97%