Merck
  • Routine Karyotyping Reveals Frequent Mosaic Reciprocal Chromosome Translocations in Swine: Prevalence, Pedigree, and Litter Size.
Routine Karyotyping Reveals Frequent Mosaic Reciprocal Chromosome Translocations in Swine: Prevalence, Pedigree, and Litter Size.
Scientific reports (2020-05-06)
Samira Rezaei, Brendan Donaldson, Daniel A F Villagomez, Tamas Revay, Nicolas Mary, Daniela A Grossi, W Allan King
ABSTRACT
In the routine commercial karyotype analysis on 5,481 boars, we identified 32 carriers of mosaic reciprocal translocations, half of which were carrying a specific recurrent translocation, mos t(7;9). An additional 7 mosaic translocations were identified through lymphocyte karyotype analysis from parents and relatives of mosaic carriers (nā€‰=ā€‰45), a control group of non-carrier boars (nā€‰=ā€‰73), and a mitogen assessment study (nā€‰=ā€‰20), bringing the total number of mosaic carriers to 39 cases. Mosaic translocations in all carriers were recognized to be confined to hematopoietic cells as no translocations were identified in fibroblasts cells of the carriers. In addition, negative impact on reproduction was not observed as the fertility of the carriers and their relatives were comparable to breed averages, and cryptic mosaicism was not detected in the family tree. This paper presents the first study of mosaic reciprocal translocations identified in swine through routine screening practices on reproductively unproven breeding boars while presenting evidence that these type of chromosome abnormalities are not associated with any affected phenotype on the carrier animals. In addition, the detection of recurrent mosaic translocations in this study may emphasize the non-random nature of mosaic rearrangements in swine and the potential role of genomic elements in their formation.
Sigma-Aldrich
Lectin from Phytolacca americana (pokeweed), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%