Merck
  • Methionine Availability in the Arthropod Intestine Is Elucidated through Identification of Vibrio cholerae Methionine Acquisition Systems.
Methionine Availability in the Arthropod Intestine Is Elucidated through Identification of Vibrio cholerae Methionine Acquisition Systems.
Applied and environmental microbiology (2020-03-30)
Audrey S Vanhove, Bat-Erdene Jugder, Daniela Barraza, Paula I Watnick
ABSTRACT
While only a subset of Vibrio cholerae strains are human diarrheal pathogens, all are aquatic organisms. In this environment, they often persist in close association with arthropods. In the intestinal lumen of the model arthropod Drosophila melanogaster, methionine and methionine sulfoxide decrease susceptibility to V. cholerae infection. In addition to its structural role in proteins, methionine participates in the methionine cycle, which carries out synthetic and regulatory methylation reactions. It is, therefore, essential for the growth of both animals and bacteria. Methionine is scarce in some environments, and the facile conversion of free methionine to methionine sulfoxide in oxidizing environments interferes with its utilization. To ensure an adequate supply of methionine, the genomes of most organisms encode multiple high-affinity uptake pathways for methionine as well as multiple methionine sulfoxide reductases, which reduce free and protein-associated methionine sulfoxide to methionine. To explore the role of methionine uptake and reduction in V. cholerae colonization of the arthropod intestine, we mutagenized the two high-affinity methionine transporters and five methionine sulfoxide reductases encoded in the V. cholerae genome. We show that MsrC is the sole methionine sulfoxide reductase active on free methionine sulfoxide. Furthermore, in the absence of methionine synthesis, high-affinity methionine uptake but not reduction is essential for V. cholerae colonization of the Drosophila intestine. These findings allow us to place a lower limit of 0.05 mM and an upper limit of 0.5 mM on the methionine concentration in the Drosophila intestine.IMPORTANCE Methionine is an essential amino acid involved in both biosynthetic and regulatory processes in the bacterial cell. To ensure an adequate supply of methionine, bacteria have evolved multiple systems to synthesize, import, and recover this amino acid. To explore the importance of methionine synthesis, transport, and recovery in any environment, all of these systems must be identified and mutagenized. Here, we have mutagenized every high-affinity methionine uptake system and methionine sulfoxide reductase encoded in the genome of the diarrheal pathogen V. cholerae We use this information to determine that high-affinity methionine uptake systems are sufficient to acquire methionine in the intestine of the model arthropod Drosophila melanogaster but are not involved in virulence and that the intestinal concentration of methionine must be between 0.05 mM and 0.5 mM.
Sigma-Aldrich
L-Methionine sulfoxide
Sigma-Aldrich
D-Methionine, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%