Merck
  • Dietary composition and yeast/microalgae combination supplementation modulate the microbial ecosystem in the caecum, colon and faeces of horses.
Dietary composition and yeast/microalgae combination supplementation modulate the microbial ecosystem in the caecum, colon and faeces of horses.
The British journal of nutrition (2019-11-07)
Pauline Grimm, Sylvie Combes, Géraldine Pascal, Laurent Cauquil, Véronique Julliand
ABSTRACT
Starchy diets can induce hindgut dysbiosis in horses. The present study evaluated the impact of a yeast (Saccharomyces cerevisiae) and microalgae (Aurantiochytrium limacinum) supplementation on caecal, colonic and faecal microbial ecosystem and on blood inflammatory parameters of horses fed high-fibre or high-starch diets. Six fistulated geldings in a 2 × 2 Latin-square design were alternatively supplemented and received during each period 100 % hay (4 weeks) followed by a 56/44 hay/barley diet (3 weeks). Caecal, colonic and faecal samples were collected 4 h after the morning meal three times per diet, at 5-d intervals, to measure bacterial composition and microbial end products. Blood was simultaneously collected for measuring inflammatory markers. The starchy diet clearly modified the microbial ecosystem in the three digestive segments, with an increase of the amylolytic function and a decrease of the fibrolytic one. However, no effect of the diet was observed on the blood parameters. When horses were supplemented, no significant change was found in lipopolysaccharides, PG-E2, serum amyloid A concentrations and complete blood count neither in cellulose-utilising, starch-utilising and lactate-utilising bacteria concentrations nor in the volatile fatty acids and lactate concentrations and pH. Under supplementation, relative abundance of Family XIII Clostridiales increased in caecum and faeces irrespective of diet and relative abundance of Veillonellaceae was higher during the hay/barley diet in colon and faeces. Most variations of faecal bacterial taxa under supplementation were not observed in the hindgut. However, all variations suggested that supplementation could increase fibrolytic function whatever the diet and limit dysbiosis when the horses' diet changed from high fibre to high starch.
Sigma-Aldrich
DL-β-Hydroxymyristic acid, ≥98%
Pricing and availability is not currently available.