Merck
  • FOXO1-mediated activation of Akt plays a critical role in vascular homeostasis.
FOXO1-mediated activation of Akt plays a critical role in vascular homeostasis.
Circulation research (2014-05-31)
Harita Dharaneeswaran, Md Ruhul Abid, Lei Yuan, Dylan Dupuis, David Beeler, Katherine C Spokes, Lauren Janes, Tracey Sciuto, Peter M Kang, Shou-Ching S Jaminet, Ann Dvorak, Marianne A Grant, Erzsébet Ravasz Regan, William C Aird
ABSTRACT
Forkhead box-O transcription factors (FoxOs) transduce a wide range of extracellular signals, resulting in changes in cell survival, cell cycle progression, and several cell type-specific responses. FoxO1 is expressed in many cell types, including endothelial cells (ECs). Previous studies have shown that Foxo1 knockout in mice results in embryonic lethality at E11 because of impaired vascular development. In contrast, somatic deletion of Foxo1 is associated with hyperproliferation of ECs. Thus, the precise role of FoxO1 in the endothelium remains enigmatic. To determine the effect of endothelial-specific knockout and overexpression of FoxO1 on vascular homeostasis. We show that EC-specific disruption of Foxo1 in mice phenocopies the full knockout. Although endothelial expression of FoxO1 rescued otherwise Foxo1-null animals, overexpression of constitutively active FoxO1 resulted in increased EC size, occlusion of capillaries, elevated peripheral resistance, heart failure, and death. Knockdown of FoxO1 in ECs resulted in marked inhibition of basal and vascular endothelial growth factor-induced Akt-mammalian target of rapamycin complex 1 (mTORC1) signaling. Our findings suggest that in mice, endothelial expression of FoxO1 is both necessary and sufficient for embryonic development. Moreover, FoxO1-mediated feedback activation of Akt maintains growth factor responsive Akt/mTORC1 activity within a homeostatic range.
Sigma-Aldrich
Anti-eNOS/NOS III Antibody, CT, Upstate®, from rabbit
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse