Merck
  • Skeletal muscle triacylglycerol hydrolysis does not influence metabolic complications of obesity.
Skeletal muscle triacylglycerol hydrolysis does not influence metabolic complications of obesity.
Diabetes (2013-07-10)
Mitch T Sitnick, Mahesh K Basantani, Lingzhi Cai, Gabriele Schoiswohl, Cynthia F Yazbeck, Giovanna Distefano, Vladimir Ritov, James P DeLany, Renate Schreiber, Donna B Stolz, Noah P Gardner, Petra C Kienesberger, Thomas Pulinilkunnil, Rudolf Zechner, Bret H Goodpaster, Paul Coen, Erin E Kershaw
ABSTRACT
Intramyocellular triacylglycerol (IMTG) accumulation is highly associated with insulin resistance and metabolic complications of obesity (lipotoxicity), whereas comparable IMTG accumulation in endurance-trained athletes is associated with insulin sensitivity (the athlete's paradox). Despite these findings, it remains unclear whether changes in IMTG accumulation and metabolism per se influence muscle-specific and systemic metabolic homeostasis and insulin responsiveness. By mediating the rate-limiting step in triacylglycerol hydrolysis, adipose triglyceride lipase (ATGL) has been proposed to influence the storage/production of deleterious as well as essential lipid metabolites. However, the physiological relevance of ATGL-mediated triacylglycerol hydrolysis in skeletal muscle remains unknown. To determine the contribution of IMTG hydrolysis to tissue-specific and systemic metabolic phenotypes in the context of obesity, we generated mice with targeted deletion or transgenic overexpression of ATGL exclusively in skeletal muscle. Despite dramatic changes in IMTG content on both chow and high-fat diets, modulation of ATGL-mediated IMTG hydrolysis did not significantly influence systemic energy, lipid, or glucose homeostasis, nor did it influence insulin responsiveness or mitochondrial function. These data argue against a role for altered IMTG accumulation and lipolysis in muscle insulin resistance and metabolic complications of obesity.
Sigma-Aldrich
Anti-phospho-Akt1/PKBα (Ser473) Antibody, clone SK703, rabbit monoclonal, clone SK703, Upstate®, from rabbit
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, Upstate®, from rabbit