Merck
  • Role of interferon-gamma and nitric oxide in pulmonary edema and death induced by lipopolysaccharide.
Role of interferon-gamma and nitric oxide in pulmonary edema and death induced by lipopolysaccharide.
American journal of respiratory and critical care medicine (2000-01-05)
H Heremans, C Dillen, M Groenen, P Matthys, A Billiau
ABSTRACT
Mice given lipopolysaccharide (LPS) intravenously developed lung edema, which was maximum after 6 h. Tumor necrosis factor, interleukin 12 (IL-12), IL-6, and interferon-gamma (IFN-gamma) appeared in the serum, and levels of nitrogen oxide (NO) derivatives were increased in serum and bronchoalveolar fluid. Mice pretreated with neutralizing anti-IFN-gamma antibodies had lower serum levels of IFN-gamma, and fewer died. However, levels of other cytokines and NO derivatives as well as lung edema were unchanged. If IFN-gamma and LPS were given together, pulmonary edema was less, but levels of cytokines and NO derivatives in serum were raised, and the mortality was greater. IFN-gamma receptor knockout mice had more edema after LPS, but were less sensitive to the lethal effects. Treatment with anti-IL-12 antibody inhibited IFN-gamma induction and reduced mortality, but had no effect on the lung edema; exogenous IL-12 also failed to affect edema, but boosted serum cytokine levels and increased the mortality. Aminoguanidine, an inhibitor of NO synthase, protected against pulmonary edema, but did not modify the lethal effects of LPS. Clearly, in this model, early pulmonary edema and lethality are not directly related, and induced IFN-gamma has no role in causing early lung edema, but augments other events that result in death.
Sigma-Aldrich
Lipopolysaccharides from Serratia marcescens, purified by phenol extraction
Pricing and availability is not currently available.