Merck
  • Improved foot-and-mouth disease vaccine, O TWN-R, protects pigs against SEA topotype virus occurred in South Korea.
Improved foot-and-mouth disease vaccine, O TWN-R, protects pigs against SEA topotype virus occurred in South Korea.
Veterinary microbiology (2019-09-11)
Joo-Hyung Choi, Mi-Kyeong Ko, Sung Ho Shin, Su-Hwa You, Hye-Eun Jo, Hyundong Jo, Min Ja Lee, Su-Mi Kim, Jong-Soo Lee, Byounghan Kim, Jong-Hyeon Park
ABSTRACT
Foot-and-mouth disease (FMD) is a highly contagious disease and causes economic damage at a national level. In particular, the type O FMD virus (FMDV) is a serotype that causes FMD outbreaks most frequently in the world. In recent years, Southeast Asia (SEA), Middle East-South Asia (ME-SA), and Cathay topotype-mediated FMD are prevalent in Asia, among which the SEA and ME-SA topotypes cause a majority of the outbreaks. The SEA topotype virus is more likely to infect both cattle and pigs simultaneously, thereby resulting in more severe damages; thus, it is necessary to study the protection ability of the candidate vaccines of this topotype after immunization. In this study, an experimental vaccine for pigs was produced using a vaccine strain that contains the structural protein of the O Taiwan97 strain, which was derived from the Cathay topotype, and its effect was evaluated. In the immunization test in pigs and cattle, the antibody titers were found to be elevated two weeks after immunization and very high titers of neutralizing antibodies were formed after four weeks. After the second inoculation, very high titers of neutralizing antibodies were produced in both species in the fourth week after immunization, and the antibodies maintained for up to six months and three months in cattle and pigs, respectively. No significant immunological difference in antibody production was observed in cattle and pigs. This study confirmed that complete protection from the challenge of the SEA topotype virus (O/Jincheon/SKR/2014), although the antibody titers against O/Jincheon/SKR/2014 strain were not that high, was achieved through immunization with the newly developed Cathay topotype vaccine in pigs.
Sigma-Aldrich
Poly(ethylene glycol), average Mn 6,000
Pricing and availability is not currently available.