Merck
Optimal Substrate-Trapping Mutants to Discover Substrates of HDAC1.
Chembiochem : a European journal of chemical biology (2019-02-01)
Inosha D Gomes, Mary Kay H Pflum
ABSTRACT
Histone deacetylase 1 (HDAC1) regulates transcription by deacetylating histones. In addition to histones, several non-histone proteins are HDAC1 substrates, which suggests a role for HDAC1 beyond epigenetics. Unfortunately, the identification of non-histone substrates has been largely serendipitous, which makes full characterization of HDAC1 functions difficult. To overcome this challenge, inactive "trapping" mutants were recently developed to identify HDAC1 substrates. To optimize substrate trapping, the relative trapping abilities of 17 inactive HDAC1 mutants was assessed. HDAC1 H141A, F150A, and C151A showed strong binding to substrates LSD1 and p53. Interestingly, each mutant preferentially trapped a different substrate. By combining several inactive mutants, the trapping strategy will facilitate the discovery of new HDAC1 substrates and shed light on the variety of HDAC1-related functions in cell biology.
Sigma-Aldrich
Anti-LSD1 (AOF2) (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)