Merck
  • Differential trafficking of albumin and IgG facilitated by the neonatal Fc receptor in podocytes in vitro and in vivo.
Differential trafficking of albumin and IgG facilitated by the neonatal Fc receptor in podocytes in vitro and in vivo.
PloS one (2019-02-28)
James Dylewski, Evgenia Dobrinskikh, Linda Lewis, Pantipa Tonsawan, Makoto Miyazaki, Parmjit S Jat, Judith Blaine
ABSTRACT
Proteinuria is strongly associated with kidney disease progression but the mechanisms underlying podocyte handling of serum proteins such as albumin and IgG remain to be elucidated. We have previously shown that albumin and IgG are transcytosed by podocytes in vitro. In other epithelial cells, the neonatal Fc receptor (FcRn) is required to salvage albumin and IgG from the degradative pathway thereby allowing these proteins to be transcytosed or recycled. Here we directly examine the role of FcRn in albumin and IgG trafficking in podocytes by studying handling of these proteins in FcRn knockout (KO) podocytes in vitro and in a podocyte-specific FcRn knockout mice in vivo. In vitro, we find that knockout of FcRn leads to IgG accumulation in podocytes but does not alter albumin trafficking. Similarly, in vivo, podocyte-specific knockout of FcRn does not result in albumin accumulation in podocytes in vivo as measured by mean albumin fluorescence intensity whereas these mice demonstrate significant intraglomerular accumulation of IgG over time. In addition we find that podocyte-specific FcRn KO mice demonstrate mesangial expansion as they age and activation of mesangial cells as demonstrated by increased expression of α-smooth muscle actin. Taken together, these results suggest that trafficking pathways for albumin and IgG differ in podocytes and that sustained disruption of trafficking of plasma proteins alters glomerular structure.
Sigma-Aldrich
Anti-Human IgG (Fc specific) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Podocin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution