Merck
  • Immobilization of invertase on sepharose-linked enzyme glycosyl recognizing polyclonal antibodies.
Immobilization of invertase on sepharose-linked enzyme glycosyl recognizing polyclonal antibodies.
Biotechnology and bioengineering (2008-07-22)
F Jafri, M Saleemuddin
ABSTRACT
Polyclonal antibodies directed against the yeast invertase glycosyls were raised by immunizing rabbits with neoglycoprotein-I and neoglycoprotein-II. The neoglycoproteins were prepared by separately coupling the N-linked large and small molecular weight yeast invertase oligosaccharides respectively to bovine serum albumin with the help of glutaraldehyde. Antibodies specifically recognizing the invertase oligosaccharides were purified from the sera of rabbits immunized with either neoglycoprotein using an affinity column of sepharose 4B-linked yeast invertase. Specific immunoaffinity supports for the immobilization of invertase were constructed by coupling the affinity-purified antineoglycoprotein-I or antineoglycoprotein-II antibodies to cyanogen bromide activated sepharose-4B. Both the affinity adsorbants were effective in binding and improving the thermal stability of invertase. (c) 1997 John Wiley & Sons, Inc. Biotechnol Bioeng 56: 605-609, 1997.
Sigma-Aldrich
Cyanogen bromide-activated-Sepharose 4 Fast Flow, lyophilized powder
Sigma-Aldrich
Cyanogen bromide-activated-Sepharose 4B, lyophilized powder