Merck
  • Immunodetection of human telomerase reverse-transcriptase (hTERT) re-appraised: nucleolin and telomerase cross paths.
Immunodetection of human telomerase reverse-transcriptase (hTERT) re-appraised: nucleolin and telomerase cross paths.
Journal of cell science (2006-06-15)
Ying-Li Wu, Charles Dudognon, Eric Nguyen, Josette Hillion, Frédéric Pendino, Ilona Tarkanyi, Janos Aradi, Michel Lanotte, Jian-Hua Tong, Guo-Qiang Chen, Evelyne Ségal-Bendirdjian
ABSTRACT
The involvement of telomerase in cellular immortalization and senescence has often been assessed by means of telomerase expression at the RNA level and quantification of telomerase activity by the telomeric repeat amplification protocol assay. However, these methods either neglected the existence of various telomerase splice variants, or ignored the nonconventional functions of telomerase independent of its ability to elongate and maintain telomere length. Immunodetection of telomerase is now being recognized as a necessary approach to precisely elucidate its roles in oncogenesis and senescence. A few antibodies directed against the catalytic subunit of the human telomerase (hTERT) are currently used but their specificity is not always demonstrated. A survey of the literature showed inconsistencies and led us to comparatively re-evaluate the most frequently used antibodies. Surprisingly, mass spectrometry, two-dimensional gel analysis and immunofluorescent experiments revealed that the most frequently used hTERT immunoprobe, a mouse monoclonal antibody that was claimed to be directed against an hTERT protein epitope, in fact recognizes nucleolin rather than telomerase. Our findings have interesting implications regarding the biology of nucleolin and telomerase in the context of pathophysiological investigations recently carried out.
Sigma-Aldrich
Anti-Heat Shock Protein 90β Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution