Merck
  • Calbindin-D28K mediates 25(OH)D3/VDR-regulated bone formation through MMP13 and DMP1.
Calbindin-D28K mediates 25(OH)D3/VDR-regulated bone formation through MMP13 and DMP1.
Journal of cellular biochemistry (2018-01-30)
Li-Ping Yang, Ya-Ping Dong, Wen-Ting Luo, Tong Zhu, Qi-Wei Li, Li-Jun Zhang, Juan Kong, Zheng-Wei Yuan, Qun Zhao
ABSTRACT
Calcium binding protein calbindin-D28K (CaBP28K) mediates the relationship between vitamin D and calcium, but its mechanism remains unclear during bone formation. The present study reports that maternal CaBP28K levels were positively correlated with paired umbilical cord CaBP28K levels. In addition, CaBP28K levels were positively correlated with the body length, and head and chest circumferences of neonates, but negatively correlated with maternal 25(OH)D3 levels. CaBP28K was also downregulated in MC3T3-E1 osteoblasts when treated with 1,25(OH)2D or VDR overexpression, but was upregulated in the femur of 1α(OH)ase(-/-) mice. Furthermore, it was found CaBP28K may influence cell differentiation and matrix formation through the regulation of DMP1 and the interaction with MMP13 in osteoblasts. This suggests that CaBP28K could be a candidate for the negative role of 1,25(OH)2D/VDR in regulating bone mass.
Sigma-Aldrich
Human CALB1 / Calbindin ELISA Kit
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)