Merck
  • Tuning Liposome Membrane Permeability by Competitive Coiled Coil Heterodimerization and Heterodimer Exchange.
Tuning Liposome Membrane Permeability by Competitive Coiled Coil Heterodimerization and Heterodimer Exchange.
Langmuir : the ACS journal of surfaces and colloids (2018-05-15)
Camilla Skyttner, Karin Enander, Christopher Aronsson, Daniel Aili
ABSTRACT
Membrane-active peptides that enable the triggered release of liposomal cargo are of great interest for the development of liposome-based drug delivery systems but require peptide-lipid membrane interactions that are highly defined and tunable. To this end, we have explored the possibility to use the competing interactions between membrane partitioning and heterodimerization and the folding of a set of four different de novo designed coiled coil peptides. Covalent conjugation of the cationic peptides triggered rapid destabilization of membrane integrity and the release of encapsulated species. The release was inhibited when introducing complementary peptides as a result of heterodimerization and folding into coiled coils. The degree of inhibition was shown to be dictated by the coiled coil peptide heterodimer dissociation constants, and liposomal release could be reactivated by a heterodimer exchange to render the membrane bound peptide free and thus membrane-active. The possibility to tune the permeability of lipid membranes using highly specific peptide-folding-dependent interactions delineates a new possible approach for the further development of responsive liposome-based drug delivery systems.
Product Description
Avanti
18:1 MPB PE, Avanti Research - A Croda Brand 870012P, powder
Avanti
18:1 MPB PE, Avanti Research - A Croda Brand 870012C
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand