Merck
  • Unveiling the important roles of coexisting contaminants on photochemical transformations of pharmaceuticals: Fibrate drugs as a case study.
Unveiling the important roles of coexisting contaminants on photochemical transformations of pharmaceuticals: Fibrate drugs as a case study.
Journal of hazardous materials (2018-07-11)
Ya-Nan Zhang, Yangjian Zhou, Jiao Qu, Jingwen Chen, Jianchen Zhao, Ying Lu, Chao Li, Qing Xie, Willie J G M Peijnenburg
ABSTRACT
Pharmaceuticals are a group of ubiquitous emerging pollutants, many of which have been shown to undergo efficient photolysis in the environment. Photochemically produced reactive intermediates (PPRIs) sensitized by the pharmaceuticals in sunlit natural waters may induce photodegradation of coexisting compounds. In this study, the roles of coexisting contaminants on the phototransformation of pharmaceuticals were unveiled with the fibrate drugs gemfibrozil (GMF), fenofibrate (FNF), and fenofibric acid (FNFA) as model compounds. GMF undergoes initial concentration dependent photodegradation due to the involvement of singlet oxygen (1O2) initiated self-sensitized photolysis, and undergoes pH dependent photodegradation due to dissociation and hydroxyl radical (OH) generation. The decarboxylated intermediates of GMF and coexisting FNFA significantly accelerated the photodegradation of GMF. The promotional effects of the decarboxylated intermediates are attributed to generation of PPRIs, e.g. 1O2, superoxide (O2-), that subsequently react with GMF. Besides, FNFA can also promote the photodegradation of GMF through the electron transfer reaction from ground state GMF to excited state FNFA, leading to the formation of decarboxylated intermediates. The formed intermediates can subsequently also facilitate GMF photodegradation. The results presented here provided valuable novel insights into the effects of coexisting contaminants on the photodegradation of pharmaceuticals in polluted waters.
Sigma-Aldrich
2,5-Dimethylanisole, 99%
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
2,5-Dimethyl-1,4-benzoquinone, ≥98.0%