Merck
  • Factors involved in specific transcription by mammalian RNA polymerase II. Identification and characterization of factor IIH.
Factors involved in specific transcription by mammalian RNA polymerase II. Identification and characterization of factor IIH.
The Journal of biological chemistry (1992-02-05)
O Flores, H Lu, D Reinberg
ABSTRACT
Two new factors required for transcription of class II genes have been identified. These factors, TFIIH and TFIIJ, were required together with the previously described general factors (TFIIA, TFIIB, TFIID, TFIIE, and TFIIF) and RNA polymerase II for transcription of different class II genes. TFIIH was extensively purified, and the activity appeared to coelute with polypeptides of 33 and 95 kDa. The role of TFIIH and TFIIJ in preinitiation complex assembly was analyzed using mobility shift assays. It was found that TFIIH and TFIIJ association with the preinitiation complex was ordered and required the previous assembly of a preinitiation complex intermediate containing factors IID, IIB, IIF, IIE, and RNA polymerase II. A model for the ordered assembly of the general factors and RNA polymerase II is presented.
Sigma-Aldrich
TFIIH, native complex human, ≥75% (SDS-PAGE)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TFIIH, p62 subunit human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Pricing and availability is not currently available.