
Vinpocetine

Synonym(s):
(3α,16α)-Eburnamenine-14-carboxylic acid ethyl ester, Eburnamenine-14-carboxylic acid ethyl ester, Vinpocetine
Empirical Formula (Hill Notation):
C22H26N2O2
CAS Number:
Molecular Weight:
350.45
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4