
Vasicinone

Synonym(s):
(3S)-2,3-Dihydro-3-hydroxypyrrolo[2,1-b]quinazolin-9(1H)-one
Empirical Formula (Hill Notation):
C11H10N2O2
CAS Number:
Molecular Weight:
202.21
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing