Skip to Content
Merck
trans-2-Heptenal ≥95%

trans-2-Heptenal

trans-2-Heptenal ≥95%
Linear Formula:
CH3(CH2)3CH=CHCHO
CAS Number:
Molecular Weight:
112.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing