
Ticagrelor for system suitability

Synonym(s):
Ticagrelor, (1S,2S,3R,5S)-3-[7-{[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino}-5-(propylsulfanyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)-1,2-cyclopentanediol
Empirical Formula (Hill Notation):
C23H28F2N6O4S
CAS Number:
Molecular Weight:
522.57
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing