
Sodium tetrathionate dihydrate

Linear Formula:
NaO3SS2SO3Na · 2H2O
CAS Number:
Molecular Weight:
306.27
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing