
Sodium fusidate

Synonym(s):
Fusidic acid sodium salt, Fucidin
Linear Formula:
C31H47O6Na
CAS Number:
Molecular Weight:
538.69
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing