
Salicylic acid impurity B

Synonym(s):
4-Hydroxyisophthalic acid, 4-Hydroxy 1,3-benzenedicarboxylic acid
Linear Formula:
HOC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
182.13
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing