
Salbutamol aldehyde impurity

Synonym(s):
5-(2-tert-Butylamino-1-hydroxyethyl)-2-hydroxybenzaldehyde, 5-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxybenzaldehyde
Empirical Formula (Hill Notation):
C13H19NO3
CAS Number:
Molecular Weight:
237.29
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing