
Ramelteon

Synonym(s):
Ramelteon solution
Empirical Formula (Hill Notation):
C16H21NO2
CAS Number:
Molecular Weight:
259.34
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing