
Quetiapine Related Compound B

Synonym(s):
Norquetiapine dihydrochloride, 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine dihydrochloride
Empirical Formula (Hill Notation):
C17H17N3S · 2HCl
CAS Number:
Molecular Weight:
368.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2